missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK1, Mouse anti-Human, Clone: 4C2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00204851-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: CTPLMVATLHPQVATITPQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYSSpecifications
| HIPK1 | |
| Monoclonal | |
| Unconjugated | |
| homeodomain interacting protein kinase 1 | |
| KIAA0630/MGC26642/MGC33446/MGC33548/Myak/Nbak2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 4C2 | |
| In 1x PBS, pH 7.4 | |
| BC028408 | |
| HIPK1 | |
| HIPK1 (AAH28408, 330 a.a. ∼ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 204851 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction