Learn More
HIPK1/HIPK2/HIPK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA5106247
Product Code. 16652685
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Antibody detects endogenous levels of total HIPK1/2/3.
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIPK1/HIPK2/HIPK3
Polyclonal
Unconjugated
HIPK3
1110014O20Rik; 1110062K04Rik; androgen receptor-interacting nuclear protein kinase; ANPK; B230339E18Rik; DYRK6; EMBL:ABO47653.1}; fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; hHIPk2; Hipk1; hipk1 {ECO:0000312; HIPK2; HIPK3; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain interacting protein kinase 2; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 1; Homeodomain-interacting protein kinase 2; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; KIAA0630; mir-1902; Myak; Nbak1; Nbak2; nbak3; nuclear body associated kinase 2b; nuclear body-associated kinase 1; Nuclear body-associated kinase 2; nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; PRO0593; protein kinase Myak; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; YAK1
Rabbit
Affinity chromatography
RUO
10114, 15257, 15258, 15259, 204851, 28996, 362342, 365895, 83617
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
A4L9P5, O88850, O88904, Q86Z02, Q9ERH7, Q9H2X6, Q9H422, Q9QZR5
HIPK1, HIPK2, HIPK3
A synthesized peptide derived from human HIPK1(Accession Q86Z02), corresponding to amino acid residues Y331-V381.
100 μL
Primary
Human, Mouse, Rat