missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIP55 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA582988
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Immunogen sequence: PPEQETFYEQ PPLVQQQGAG SEHIDHHIQG QGLSGQGLCA RALYDYQAAD DTEISFDPEN LITGIEVIDE GWWRGYGPDG H Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 86%.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
Specifica
| HIP55 | |
| Polyclonal | |
| Unconjugated | |
| DBNL | |
| Abp1; Actin-binding protein 1; cervical mucin-associated protein; Cervical SH3P7; CMAP; DBNL; dbnl {ECO:0000312; drebrin F; drebrin like; drebrin-F; drebrin-like; drebrin-like protein; EMBL:AAH72483.1}; HIP55; HIP-55; HPK1-interacting protein of 55 kDa; mAbp1; PP5423; SH3 domain-containing protein 7; Sh3p7; src homology 3 domain-containing protein HIP-55 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13169, 28988, 83527 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q62418, Q9JHL4, Q9UJU6 | |
| DBNL | |
| Recombinant protein corresponding to Human DBNL. Recombinant protein control fragment (Product #RP-92734). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering