missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIP1R Monoclonal Antibody (K1E009_9H2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555935
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: DRDLQIESLK REVEMLRSEL EKIKLEAQRY IAQLKSQVNA LEGELEEQRK QKQKALVDNE QLRHELAQLR AAQLEGERSQ GLREEAERKA SATEARYNKL KEKHSELVHV HAELLRKNAD TAKQLTVTQQ SQEEVARVKE QLAFQVEQVK RESELKLEEK SDQLEKLKRE LEAKAGELAR AQEALSHTEQ
Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
Specifications
| HIP1R | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O75146 | |
| HIP1R | |
| Recombinant human HIP1R (immunogen range is 351-540). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K1E009_9H2 | |
| Unconjugated | |
| HIP1R | |
| AA410023; HIP12; HIP-12; Hip1r; HIP1-related protein; HIP3; huntingtin interacting protein 1 related; huntingtin interacting protein 12; Huntingtin-interacting protein 12; huntingtin-interacting protein 1-related protein; ILWEQ; KIAA0655; mKIAA0655 | |
| Mouse | |
| Protein A | |
| RUO | |
| 9026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction