missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP14/ZDHHC17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£122.57 - £303.85
Specifications
| Antigen | HIP14/ZDHHC17 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.Specifications
| HIP14/ZDHHC17 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| E9PTT0, Q80TN5, Q8IUH5 | |
| 23390, 320150, 366889 | |
| HIP14; ZDHHC17 Fusion Protein Ag7763 | |
| Primary | |
| -20°C | |
| ZDHHC17 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DHHC 17, HIP 14, HIP 3, HIP14, HIP3, HSPC294, Huntingtin yeast partner H, HYPH, KIAA0946, Palmitoyltransferase ZDHHC17, ZDHHC17 | |
| Zdhhc17 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title