missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Related Antibody (1E1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB300-206
This item is not returnable.
View return policy
Description
HIP1 Related Monoclonal specifically detects HIP1 Related in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| HIP1 Related | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| FLJ14000, HIP-12, HIP12HIP1-related protein, HIP3, huntingtin interacting protein 1 related, huntingtin interacting protein 12, Huntingtin-interacting protein 12, huntingtin-interacting protein 1-related protein, ILWEQ, KIAA0655FLJ27022, MGC47513 | |
| Mouse | |
| 120 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 9026 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 10 | |
| Western Blot 1:1000, Immunohistochemistry reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin | |
| O75146 | |
| HIP1R | |
| Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt# O75146] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bacteria | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction