missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP-55 Antibody (OTI6C9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71841C
This item is not returnable.
View return policy
Description
HIP-55 Monoclonal specifically detects HIP-55 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| HIP-55 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DBNL | |
| Full length recombinant protein of human HIP-55 (NP_054782) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI6C9 | |
| Immunohistochemistry | |
| ABP1, Cervical SH3P7, CMAP, Drebrin-F, drebrin-like, drebrin-like protein, HIP-55SH3P7Cervical mucin-associated protein, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, src homology 3 domain-containing protein HIP-55 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 28988.0 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido