missing translation for 'onlineSavingsMsg'
Learn More

HINT2 Antibody (AT5B10), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18414293 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414293 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414293 Supplier Novus Biologicals Supplier No. NBP259387AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HINT2 Monoclonal antibody specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HINT2
Applications Western Blot, ELISA
Classification Monoclonal
Clone AT5B10
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2
Host Species Mouse
Immunogen Recombinant human HINT2 (18-163aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84681
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.