missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HINT1 Recombinant Rabbit Monoclonal Antibody (23GB5985), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552965
This item is not returnable.
View return policy
Description
Histidine triad nucleotide-binding protein 1 (HINT1) is a member of the histidine triad (HIT) protein family, a group of small nucleotide-binding and -hydrolyzing proteins. HINT1 interacts with several diverse proteins and has been suggested to have tumor suppressive activities. HINT1 catalyzes the hydrolysis of adenosine 5'-monophoramidate substrates such as AMP-morpholidate, but its enzymatic function does not appear to play a role in its tumor suppression. Recent experiments demonstrate that HINT1 forms a complex with POSH and JNK in vivo, inhibiting AP-1 activity and the phosphorylation of c-Jun, and this action could contribute to the tumor suppressor activity of HINT1. Other studies raise the possibility of HINT1 as a candidate gene for schizophrenia.
Specifications
| HINT1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Hint1 | |
| 17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15254, 3094, 690660 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5985 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49773, P62959, P70349 | |
| Hint1 | |
| A synthesized peptide derived from human HINT1 (1-32AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction