missing translation for 'onlineSavingsMsg'
Learn More

HINT1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18410465 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410465 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410465 Supplier Novus Biologicals Supplier No. NBP176944MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HINT1 Polyclonal antibody specifically detects HINT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HINT1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Adenosine 5'-monophosphoramidase, EC 3.-, HINT, histidine triad nucleotide binding protein 1, histidine triad nucleotide-binding protein, histidine triad nucleotide-binding protein 1, PKCI1, PKCI-1FLJ30414, PRKCNH1FLJ32340, Protein kinase C inhibitor 1, Protein kinase C-interacting protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of the human HINT1. The immunogen is located within the last 50 amino acids of HINT1. Amino Acid Squence: NKGYRMVVNEGSDGGQS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3094
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.