missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HINT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | HINT1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HINT1 Polyclonal specifically detects HINT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| HINT1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_005331 | |
| 3094 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of the human HINT1. The immunogen is located within the last 50 amino acids of HINT1. Amino Acid Squence: NKGYRMVVNEGSDGGQS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Adenosine 5'-monophosphoramidase, EC 3.-, HINT, histidine triad nucleotide binding protein 1, histidine triad nucleotide-binding protein, histidine triad nucleotide-binding protein 1, PKCI1, PKCI-1FLJ30414, PRKCNH1FLJ32340, Protein kinase C inhibitor 1, Protein kinase C-interacting protein 1 | |
| HINT1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title