missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIN-1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £385.00
Specifications
| Antigen | HIN-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17703314
|
Invitrogen™
PA5121912 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17793304
![]() |
Invitrogen™
PA5121911 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potential growth inhibitory cytokine. Classified as a secretoglobin; may have a role in airway function.Specifications
| HIN-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| Q96QR1 | |
| 100515999, 92304 | |
| Phe21-Gly104 (Accession Q96QR1), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SCGB3A1 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2 | |
| SCGB3A1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title