missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Required equipment: GM-HILED-RGB or GM-HILED-RIR
- Bag of parts to be fitted by professional
- Works with GBOX mini only
Specifications
Specifications
| Product Type | Lighting |
| Color | Far Red |
| For Use With (Equipment) | GM-HILED-RGB or GM-HILED-RIR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?