Learn More
Cytiva High Purity rNTP Solutions
Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Lifescience™ High Purity rNTP Solutions provide easy-to-use, time-saving solutions.
Brand: Cytiva 27-2056-01
Code : GH
Additional Details : Weight : 0.01100kg
Description
- Each lot is tested for ribonuclease contamination to ensure success in your application
- High purity: >98% triphosphate
Specifications
Liquid | |
7.5 | |
100 mM | |
25 μmol | |
in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling |
Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity. | |
ATP, 100 mM solution | |
≥98% triphosphate | |
Colorless |