Learn More
UVP High-Performance Uv Variable Intensity Transilluminator
Product Code. 12813938
Change view
Click to view available options
Wavelength:
254 nm
Unit Size:
Each
Product Code. Wavelength unitSize
12813938 254 nm Each
1 options
Product Code. 12813938 Supplier UVP Supplier No. 95042802
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
UVP's high performance transilluminators feature 25W tubes and a specially constructed filter that provides the most uniform light emission possible from a tube based unit.
The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. High, medium and low intensity settings make the unit suitable for both visualisation and band cutting. An internal fan improves the tube lifetime.Dimensions: 337mm x 486mm x 143mm
Specifications
Wavelength 254 nm