missing translation for 'onlineSavingsMsg'
Learn More

ANSELL™ High Performance Reusable Food-approved AlphaTec™ Gloves

Product Code. 17923542 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17923542 9 12 pairs
17983572 8 12 pairs
17903542 7 12 pairs
17973572 10 12 pairs
17993572 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17923542 Supplier ANSELL™ Supplier No. 58010090

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Durable yet ergonomic 0.28 mm nitrile glove.

  • Chemical glove with all day comfort which strikes a fantastic balance between value and durability
  • 11 mil or 0.28 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offering protection against light acids, greases, oils and liquids
  • Engineered fit to allow optimum balance of fit and flexibility both on its own or as an over glove
  • Long cuff extended protection to the forearm
  • Silicone and latex free
TRUSTED_SUSTAINABILITY

Tekniske data

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) ANSI ABR 4|EN 388:2016 +A1:2018|EN 421:2010|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Food Contact
Chlorinated Yes
Applications Industrial
Color Blue
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.28 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.28 mm
Product Type Chemical-resistant Gloves
Size 9
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Raised Diamond
Lining Unlined
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.