missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, High Nickel, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, High Nickel, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Copper/Phosphorus/Silicon/Manganese/Cobalt/Chromium/Nickel/Aluminium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.035 %/Copper 0.0363 %/Phosphorus 0.04 %/Silicon 0.255 %/Manganese 0.303 %/Cobalt 0.4 %/Chromium 0.409 %/Nickel 31.47 %/Aluminium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction