missing translation for 'onlineSavingsMsg'
Learn More

High Level ICV Stock SS, Ca, Fe, K, Mg, Na @ 2000 mg/l, SECOND SOURCE, 500 mL**1600601** , VHG™

Product Code. 17910886
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910886 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910886 Supplier VHG Labs Supplier No. VHG1600601500
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

High Level ICV Stock SS, Ca, Fe, K, Mg, Na @ 2000 mg/l, SECOND SOURCE, 500 mL**1600601**
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Iron/Magnesium/Potassium/Sodium
CAS 7440-70-2, 7439-89-6, 7439-95-4, 9-7-7440, 7440-23-5
Methods ICP/ICP-MS
Analyte or Component Names Calcium/Iron/Magnesium/Potassium/Sodium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Calcium 2000 mg/L/Iron 2000 mg/L/Magnesium 2000 mg/L/Potassium 2000 mg/L/Sodium 2000 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.