missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ High-Intensity Perfusion CHO Medium

Product Code. 30283494 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 L
15 L
100 L
370 L
Unit Size:
15 L
2 L
Each
This item is not returnable. View return policy

Product Code. 30283494

Brand: Gibco™ A4230201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco High-Intensity Perfusion (HIP) CHO Medium is an innovative, chemically defined, animal origin-free (AOF), cGMP medium.

Gibco High-Intensity Perfusion (HIP) CHO Medium is an innovative, chemically defined, animal origin-free (AOF), cGMP medium. This easy-to-use perfusion medium helps enable high titers in numerous CHO cell lines from the beginning to the end of the process regardless of the type of perfusion platform, all while using less medium.

Features of HIP CHO Medium include:

  • Tested with CHO-S, CHO-DG44, CHO-GS, and CHO-K1 cell lines
  • Supports high cell density of over 100 million cells/mL at 1 vessel volume/day (VVD) continuous perfusion
  • Demonstrated production of up to 1.7 g/L/day at 1 VVD continuous perfusion

Flexibility

HIP CHO Medium can be used at multiple concentrations to support seed train to production with one product. The flexible reconstitution of the medium allows dynamic tailoring of medium concentration to meet process and clone needs. Just as it can be used at multiple concentrations, HIP CHO Medium can be used in multiple perfusion processes: N-1 and seed train, intensified fed-batch, concentrated fed-batch, and continuous perfusion.In addition, HIP CHO Medium supports high cell densities at low medium exchange rates (as low as 1 VVD), which improves process efficiency.

HIP CHO Medium can help enable your move toward perfusion processes, which in turn can lead to a more compact and efficient workflow while improving productivity and quality.

Applications of HIP CHO Medium:

  • Suspension CHO cells
  • Selection system such as glutamine synthetase (GS) and dihydrofolate reductase (DHFR)
  • Passaging, seed train, and protein production applications
  • Optimized for use without feeds in perfusion application (other than glucose)
TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO-S, CHO-GS, CHO-K1, CHO-DG44
Concentration 34.92 g/L
Form Dry Powder
Product Type Perfusion CHO Medium
Sterility Non-sterile
With Additives Glucose
Without Additives No Glutamine
Manufacturing Quality cGMP
Quantity 2 L
Shipping Condition Wet Ice
Format Securitainer
Reconstituted Volume 2 L
Content And Storage Dry format medium. Store in the dark and dry at 2–8°C.

Culture Type Suspension Cell Culture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.