Learn More
Kartell™ High Impact Polystyrene Balance Boats
Provide an easy and inexpensive system for weighing any medium.
£79.20 - £324.00
Product Code | Brand | Color | Capacity (Metric) | Shape | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Capacity (Metric) | Shape | Dimensions (L x W x H) | Price | Quantity & Availability | ||
10447512
|
Kartell™
1110003 |
White | 5 mL | Diamond | 31 x 53 x 5.3 mm |
£148.00
Pack of 1000 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
12627805
|
Kartell™
1110300 |
White | 7 mL | Square | 43 x 43 x 8 mm |
£212.00
Pack of 1000 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10662163
|
Kartell™
1110103 |
White | 30 mL | Diamond | 56 x 85 x 14.5 mm |
£149.00
Pack of 500 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||
10192122
|
Kartell™
1110109 |
Black | 30 mL | Diamond | 56 x 85 x 14.5 mm |
£79.20
Pack of 500 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10738294
|
Kartell™
1110203 |
White | 100 mL | Diamond | 96 x 134.5 x 18.5 mm |
£104.00
Pack of 250 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10345812
|
Kartell™
1110209 |
Black | 100 mL | Diamond | 96 x 134.5 x 18.5 mm |
£103.00
Pack of 250 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10736962
|
Kartell™
1110400 |
White | 100 mL | Square | 78.5 x 78.5 x 23 mm |
£236.00
Pack of 1000 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10325781
|
Kartell™
1110500 |
White | 250 mL | Square | 134 x 134 x 23 mm |
£324.00
Pack of 1000 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||
10325962
|
Kartell™
1110009 |
Black | 5 mL | Diamond | 31 x 55 x 4.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Provide an easy and inexpensive system for weighing any medium.
- Lightweight weighing boats
- Non-contaminating (made of HIPS with hydrophobic surface)
- Easily fashioned to make funnel shape
- Available in two models: diamond (DS) and square (SQ)
- DS model available in white or black for visibility of trace sample remains adhering to boat
- Reproducible tare weight: batch to batch variation less than 5%
- Antistatic
Specifications
Weighing Boat |