missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Breathability with durability
- Cloth-like, microporous material
- General purpose protection from dirt, grime, splashes and spills
- Economical and lightweight
- Low linting and antistatic
- Elastic wrists, five-snap closure and two pockets
- White
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | Microporous Film |
| Color | White |
| Closure Type | Snaps |
| No. of Pockets | 2 |
| For Use With (Application) | General purpose protection from dirt, grime, splashes, and spills |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?