missing translation for 'onlineSavingsMsg'
Learn More

Brady™ High Density Thermal Transfer Printer Ribbon

Product Code. 11865245
Change view
Click to view available options
Color:
Blue
Green
Magenta
Red
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11865245 Magenta Each
11889504 Blue Each
11860845 Green Each
11870845 Red Each
11880845 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11865245 Supplier Brady™ Supplier No. 114554

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High Density Thermal Transfer Printer Ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.

  • High density ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.

Specifications

Product Type Printer Ribbon
Color Magenta
For Use With (Equipment) BMP71 Label Printer
Length (English) 50 yd.
Width (English) 1.97 in.
Length (Metric) 45.72 m
Width (Metric) 50 mm
Dimensions (L x W) 46 m x 50.8 mm
Description Magenta 6900 Series Thermal Transfer Printer Ribbon for BMP71 Printer
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.