missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz High-Density Polyethylene Waste Bags

Product Code. p-200111933
Click to view available options
Capacity (English):
1.32 gal.
5.81 gal.
7.04 gal.
19.02 gal.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30158361

Brand: Heinz Herenz 1131738

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-density waste bags, yellow with biohazard-symbol, warning instruction and sterilization indicator patch. Autoclavable.

  • Waste bags
  • Yellow for easy identification
  • For the disposal of contaminated labware
  • Rectangle white indicator patch field turns into dark when autoclaved
  • With BIOHAZARD symbol and safety imprint in English, German, French, and Spanish languages
  • Autoclavable at 121 °C for 20 minutes (the bag must not be completely closed)
  • Thickness 50 μm (0.05 mm)
  • HD-PE

Specifications

Capacity (Metric) 22 l
Capacity (English) 5.81 gal.
Color Yellow
Disposable Single-use
Sterility Non-sterile
Autoclavable Autoclavable
Material HDPE
Product Type Biohazard Waste Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.