missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ High Content Assay Buffers and Accessory Reagents

Product Code. 10319674 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10319674 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10319674 Supplier Thermo Scientific™ Supplier No. 8408400

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform high content screening, analysis and cell biology experiments using the buffer kits, standalone buffers and supplemental reagents.

Perform high content screening, analysis and cell biology experiments using the buffer kits, standalone buffers and supplemental reagents using Thermo Scientific™ Buffers and Accessory Reagents.

Our portfolio of buffers and accessory reagents for high content and cell biology applications include standard wash, permeabilization and blocking buffers, neurite outgrowth buffer, BrdU, MgCl2 and goat serum. Standard buffers are provided as kits for ease-of-use or standalone components with larger volumes for affordability.

Available Buffers and Accessory Reagents:

  • Buffer Kt 1 aka B1 (8421000)
  • Buffer Kit 2 aka B2 (8422000)
  • Buffer Kit 3 aka B3 (8423000)
  • Blocking Buffer (8408500)
  • BrdU (8419200)
  • MgCl2 (8419300)
  • Neurite Outgrowth Buffer (8406900)
  • Permeabilization Buffer (8408400)
  • Wash Buffer (8408200)
  • Wash Buffer II (8408300)

Ordering Information:

Volumes of buffers in kits are different than standalone buffers. Kits contain: wash buffers, permeabilization buffer, blocking buffer and DAPI/Hoechst dye.

TRUSTED_SUSTAINABILITY

Specifications

Description Cellomics Permeabilization Buffer (10X)
Quantity 500 mL
Formulation 10X Dulbecco's PBS with 1% Triton X-100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.