missing translation for 'onlineSavingsMsg'
Learn More

Manutan High back chair, heavy duty, black

Product Code. 12885168
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12885168

Brand: Manutan A331114

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High back for maximum back and shoulder comfort. Fully upholstered in hard-wearing fabric with integral lumbar support and adjustable arms. Heavy duty asynchro mechanism for independent seat and back adjustment with adjustable arms included. Meets BS 5459 safety requirements.

  • Black
  • Back dimensions (H x W) : 730mm x 530mm
  • Seat dimensions (W x D) : 530mm x 490mm
  • Seat height : 490mm to 550mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.