missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIGD2A Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21440C
This item is not returnable.
View return policy
Description
HIGD2A Polyclonal antibody specifically detects HIGD2A in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HIGD2A | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| HIG1 domain family member 2A, HIG1 domain family, member 2A, HIG1 hypoxia inducible domain family, member 2A, MGC2198 | |
| Produced in rabbits immunized with E. coli-derived Human HIGD2A fragment | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 192286 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction