missing translation for 'onlineSavingsMsg'
Learn More

HIGD1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18339744 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339744 25 μg 25µL
18362093 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18339744 Supplier Novus Biologicals Supplier No. NBP31716125UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIGD1B Polyclonal antibody specifically detects HIGD1B in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD1B
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias CLST 11240 protein, CLST 11240-15 protein, CLST11240, CLST11240-15, HIG1 domain family member 1B, HIG1 domain family, member 1B, HIG1 hypoxia inducible domain family, member 1B, Protein CLST 11240
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: AVYTMYSDYVKRMAQDAGEK
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51751
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.