missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIGD1A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | HIGD1A |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345444
|
Novus Biologicals
NBP3-03930-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18382437
|
Novus Biologicals
NBP3-03930-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIGD1A Polyclonal specifically detects HIGD1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HIGD1A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 25994 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 7420700H20Rik, DKFZP564K247, HIG1 domain family member 1A, HIG1 domain family, member 1A, HIG1 hypoxia inducible domain family, member 1A, HIG1DKFZp564K247, hypoxia inducible gene 1,2210020B17Rik, Hypoxia-inducible gene 1 protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1-93 of human HIGD1A (NP_054775.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title