missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIGD1A Polyclonal specifically detects HIGD1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | HIGD1A |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | 7420700H20Rik, DKFZP564K247, HIG1 domain family member 1A, HIG1 domain family, member 1A, HIG1 hypoxia inducible domain family, member 1A, HIG1DKFZp564K247, hypoxia inducible gene 1,2210020B17Rik, Hypoxia-inducible gene 1 protein |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-93 of human HIGD1A (NP_054775.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?