Learn More
Invitrogen™ HIGD1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589915
Description
Immunogen sequence: MSTDTGVSLP SYEEDQGSKL IRKAKEAPFV PVGIAGFAAI VAYGLYKLKS RGNTKMSIHL IHMRVAAQGF VVGAMTVGMG YSMYREFWAK PKP; Positive Samples: HeLa, HepG2, 293T, HepG2, K-562; Cellular Location: Mitochondrion inner membrane, Mitochondrion membrane, Multi-pass membrane protein.
Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
Specifications
| HIGD1A | |
| Polyclonal | |
| Unconjugated | |
| HIGD1A | |
| 2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; Hig1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25994 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9Y241 | |
| HIGD1A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-93 of human HIGD1A (NP_0547752). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.