missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIGD1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572706
This item is not returnable.
View return policy
Description
HIGD1A Polyclonal Antibody for Western Blot, ICC/IF, IHC, ELISA
Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
Specifications
| HIGD1A | |
| Polyclonal | |
| Unconjugated | |
| HIGD1A | |
| 2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; Hig1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9Y241 | |
| HIGD1A | |
| HIG1 antibody for a 19 amino acid synthetic peptide near the amino terminus of human HIG1. The immunogen is within the first 50 amino acids of HIG1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction