missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF3A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27650-1-AP-150UL
This item is not returnable.
View return policy
Description
Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.Specifications
| HIF3A | |
| Polyclonal | |
| Unconjugated | |
| HIF3A | |
| bHLHe17, HIF 3 alpha, HIF 3A, HIF 3A2, HIF 3A4, HIF3 alpha, HIF3 alpha 1, HIF3A, Inhibitory PAS domain protein, IPAS, Member of PAS protein 7, MOP7, PASD7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64344 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y2N7 | |
| HIF3A | |
| HIF3A Fusion Protein Ag26574 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction