missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIF3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536531
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 72 kDa. Purity is >95% by SDS-PAGE.
Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.
Specifications
| HIF3A | |
| Polyclonal | |
| Unconjugated | |
| HIF3A | |
| Basic-helix-loop-helix-PAS protein MOP7; BHLHE17; Class E basic helix-loop-helix protein 17; HIF-3 alpha; HIF3 alpha 1; HIF3A; HIF-3A; HIF-3-alpha; HIF3-alpha; HIF3-alpha-1; hypoxia inducible factor 3 alpha; hypoxia inducible factor 3 alpha subunit; hypoxia inducible factor 3, alpha subunit; hypoxia inducible factor 3a; hypoxia inducible factor three alpha; hypoxia-inducible factor 3 alpha; hypoxia-inducible factor 3-alpha; Inhibitory PAS domain protein; IPAS; Member of PAS protein 7; Mop7; Neonatal and embryonic PAS protein; NEPAS; PAS domain-containing protein 7; PASD7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53417, 64344, 64345 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0VBL6, Q9JHS2, Q9Y2N7 | |
| HIF3A | |
| Synthetic peptide corresponding to amino acids 323-376 of human HIF-3 alpha. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction