missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIF1AN Recombinant Rabbit Monoclonal Antibody (JG37-66)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA5-34764
This item is not returnable.
View return policy
Description
Positive Control: A549, LOVO, SiHa, human prostate cancer tissue.
Hydroxylates HIF-1 alpha at 'Asn-803' in the C-terminal transactivation domain (CAD). Functions as an oxygen sensor and, under normoxic conditions, the hydroxylation prevents interaction of HIF-1 with transcriptional coactivators including Cbp/p300-interacting transactivator. Involved in transcriptional repression through interaction with HIF1A, VHL and histone deacetylases. Hydroxylates specific Asn residues within ankyrin repeat domains (ARD) of NFKB1, NFKBIA, NOTCH1, ASB4, PPP1R12A and several other ARD-containing proteins. Also hydroxylates Asp and His residues within ARDs of ANK1 and TNKS2, respectively. Negatively regulates NOTCH1 activity, accelerating myogenic differentiation. Positively regulates ASB4 activity, promoting vascular differentiation. [UniProt].
Specifications
| HIF1AN | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NWT6 | |
| HIF1AN | |
| Synthetic peptide within Human HIF1AN aa 1-50. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JG37-66 | |
| Unconjugated | |
| HIF1AN | |
| 2310046M24Rik; A830014H24Rik; DKFZp762F1811; factor inhibiting HIF1; factor inhibiting HIF-1; FIH; FIH 1; FIH1; FIH-1; FLJ20615; FLJ22027; Hif1an; hypoxia inducible factor 1 alpha subunit inhibitor; hypoxia inducible factor 1 subunit alpha inhibitor; hypoxia inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1-alpha inhibitor; Hypoxia-inducible factor asparagine hydroxylase; peptide-aspartate beta-dioxygenase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55662 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction