missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1a Mouse anti-Human, Clone: 1H3C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66730-1-IG-20UL
This item is not returnable.
View return policy
Description
HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.Specifications
| HIF1a | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16665 | |
| HIF1A | |
| HIF1a Fusion Protein Ag15198 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 1H3C12 | |
| Unconjugated | |
| HIF1A | |
| ARNT interacting protein, bHLHe78, HIF 1 alpha, HIF 1alpha, HIF1, HIF1 ALPHA, HIF1A, Member of PAS protein 1, MOP1, PASD8 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3091 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction