missing translation for 'onlineSavingsMsg'
Learn More

HIF-2α-IN-1, MedChemExpress

Product Code. p-200098630
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16487978

Brand: Medchem Express HY19949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HIF-2(alpha)-IN-1 is a HIF-2(alpha) inhibitor has an IC50 of less than 500 nM in HIF-2(alpha) scintillation proximity assay.

HIF-2(alpha)-IN-1 is a HIF-2(alpha) inhibitor has an IC50 of less than 500 nM in HIF-2(alpha) scintillation proximity assay.

Specifications

Chemical Name or Material HIF-2α-IN-1
CAS 1799948-06-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H8F5NO4S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (246.73 mM; Need ultrasonic)
SMILES N#CC1=CC(F)=CC(OC2=CC=C3C([C@@H](O)C(F)(F)S3(=O)=O)=C2C(F)F)=C1
Molecular Weight (g/mol) 405.3
Formula Weight 405.3
Percent Purity 98.64%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.