missing translation for 'onlineSavingsMsg'
Learn More

HIF-2 alpha/EPAS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£240.00 - £375.00

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18220593
View Documents
Novus Biologicals
NBP2-58653
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18605247
View Documents
Novus Biologicals
NBP2-58653-25ul
25 μL
£240.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

HIF-2 alpha/EPAS1
Polyclonal
Rabbit
Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha,
EPAS1
IgG
Affinity Purified
96.5 kDa
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
2034
This HIF-2 alpha/EPAS1 Antibody was developed against a recombinant protein corresponding to the following amino acid sequence: GTVIYNPRNLQPQCIMCVNYVLSEIEKNDVVFSMDQTESLFKPHLMAMNSIFDSSGKGAVSEKSNFLFTKLKEEPEELAQLAPTPGDAIISLDFG
Primary
The specificity of this HIF-2 alpha/EPAS1 Antibody was verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.