missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-2 alpha/EPAS1 Antibody (ep190b) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-132CL1
This item is not returnable.
View return policy
Description
HIF-2 alpha/EPAS1 Monoclonal antibody specifically detects HIF-2 alpha/EPAS1 in Bovine,Hamster,Human,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Gel Super Shift Assays,In vivo Assay,Simple Western,Gel Supershift Assay,ChIP assay (ChIP),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| HIF-2 alpha/EPAS1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
| The immunogen recognized by this HIF-2 alpha/EPAS1 Antibody (ep190b) maps to a region between amino acids 535-631. [UniProt# Q99814] | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 2034 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Supershift, In vivo Assay, Simple Western, Gel Supershift, ChIP Assay, KnockDown, Immunocytochemistry/Immunofluorescence | |
| ep190b | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Hamster, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction