missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-2 alpha/EPAS1 Antibody (6A9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26600-100ul
This item is not returnable.
View return policy
Description
HIF-2 alpha/EPAS1 Monoclonal antibody specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications
| HIF-2 alpha/EPAS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry | |
| 6A9 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
| A synthesized peptide derived from Human HIF-2 alpha/EPAS1 [UniProt Q99814] | |
| 100 μL | |
| Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators | |
| 2034 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction