missing translation for 'onlineSavingsMsg'
Learn More

HIF-2 alpha/EPAS1 Antibody (2444A) [PerCP], Novus Biologicals™

Product Code. 30737097 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737097 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737097 Supplier Novus Biologicals Supplier No. FAB2997C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HIF-2 alpha/EPAS1 Monoclonal antibody specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2444A
Conjugate PerCP
Formulation PBS
Gene Alias Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2
Host Species Rabbit
Immunogen E. coli-derived recombinant human HIF-2 alpha/EPAS1, Ser543-Thr870, Accession # Q99814
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.