missing translation for 'onlineSavingsMsg'
Learn More

HIF-2 alpha/EPAS1 Antibody (2444A), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18342310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342310 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342310 Supplier Novus Biologicals Supplier No. NBP276454AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2444A
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha,
Gene Symbols EPAS1
Host Species Rabbit
Immunogen This recombinant HIF-2 alpha/EPAS1 Antibody (2444A) is made to a partial recombinant human HIF-2 protein (amino acids 543-870) [UniProt Q99814]
Quantity 0.1 mL
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.