missing translation for 'onlineSavingsMsg'
Learn More

HIF-1α (Hypoxia-Inducible Factor-1α) Ab-4, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ HIF-1α (Hypoxia-Inducible Factor-1α) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-1164-P0

 View more versions of this product

Product Code. 12659166

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

HIF-1 (hypoxia-inducible factor-1) is a heterodimer consisting of an alpha and a β subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF-1 is a key component of a widely operative transcriptional response activated by hypoxia, cobaltous ions, and iron chelation. HIF-1 activates transcription of hypoxia-inducible genes, including those encoding: erythropoietin, vascular endothelial growth (VEGF), heme oxygenase-1, inducible nitric oxide synthase, and the glycolytic enzymes aldolase A, enolase 1, lactate dehydrogenase A, phosphofructokinase I, and phosphoglycerate kinase 1. The C-terminal of HIF-1 alpha binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF.

Host Species: Mouse

Clone: H1α67

Isotype: IgG2b/κ

Species Reactivity: Human, Sheep, Mouse, Rat and Ferret. Others not known.

Epitope: aa 432-528

Immunogen: Recombinant hHIF-1α (aa 432-528) protein

Molecular Weight: 120kDa

Positive Control: Mammalian cells cultured under reduced O2 tension

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIF-1alpha (Hypoxia-Inducible Factor-1alpha) Ab-4
Monoclonal
200μg/mL
Mouse
100 μL
Transcription Regulation
Ferret, Human, Mouse, Rat, Sheep
IgG2b κ
Immunohistochemistry (Paraffin)
H1alpha67
Unconjugated
Recombinant hHIF-1alpha (aa 432-528) protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.