Learn More
Invitrogen™ HIF1A Human ELISA Kit
Contains sufficient reagents and buffers to assay HIF-1 alpha (hypoxia-inducible factor 1-alpha) levels in serum, plasma (EDTA, heparin, sodium citrate), and culture lysate
Brand: Invitrogen™ EHIF1A5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The Human HIF1A ELISA quantitates Hu HIF1A in human serum, plasma, or cell culture lysates. The assay will exclusively recognize both natural and recombinant Hu HIF1A. Principle of the method The Human HIF1A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.Specifications
Q16665 | |
30 pg/mL | |
480 | |
Colorimetric | |
ELISA Kit | |
Cell Lysates, Plasma, Serum | |
TMB | |
Mouse, Rat | |
Colorimetric Microplate Reader | |
HIF-1-alpha,HIF-1A,HIF-1alpha,HIF1,HIF1-ALPHA,MOP1,PASD8,bHLHe78 | |
<10% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Anti-HIF-1A precoated 96-well strip plate, lyophilized recombinant HIF-1A standard, standard diluent, 30X wash buffer, biotinylated antibody reagent, streptavidin-HRP reagent, HRP dilution buffer, TMB substrate, stop solution, contains 0.16M sulfuric acid | |
Absorbance at 450nm minus absorbance at 550nm | |
Intra-assay CV <10%; inter-assay CV <10% | |
RUO | |
Cell Lysate,50 μL; Plasma, 50 μL; Serum, 50 μL | |
Human | |
4 hr. |
81.92-20,000 pg/mL | |
HIF-1 Alpha ELISA Kit, 480 Assays, Human Species, Immunology Research Category, Colorimetric Detection Method, Colorimetric Elisa (TMB Substrate) in Clear 96-Well Strip Plate, The Complete Kits Contain Precoated 96-Well Strip Plate(S) and Lot-Optimized Re | |
Biotin | |
Colorimetric ELISA (TMB substrate) with Plates | |
Transcription factors | |
Specific to human HIF-1A and shows cross-reactivity with mouse and rat HIF-1A | |
480 Assay | |
Protein assays and analysis | |
3091 | |
Human | |
<10% | |
HRP | |
450 to 550nm | |
5 x 96 Tests | |
Immunology | |
2°C to 8°C | |
1 hr. 20 min. |