missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
- 1.NB100-105: HIF-1 alpha Antibody (H1alpha67)
- 2.NB100-131: HIF-1 alpha Antibody (ESEE122)
- 3.NB100-449: HIF-1 alpha Antibody
- 4.NB100-479: HIF-1 alpha Antibody
- 5.NB100-654: HIF-1 alpha Antibody
Specifications
Specifications
| Antigen | HIF-1 alpha |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp |
| Gene Symbols | HIF1A |
| Immunogen | For NB 100-654: Human HIF-1 alpha amino acids 432-528., For NB 100-105: Human HIF-1 alpha amino acids 432-528., For NB 100-131: Human HIF-1 alpha amino acids 329-530., For NB 100-449: Human HIF-1 alpha amino acids 775-826., For NB 100-479: Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221]. |
| Quantity | 5 Vials |
| Research Discipline | Cancer, HIF Target Genes, Hypoxia, mTOR Pathway |
| Gene ID (Entrez) | 3091 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?