missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
- 1.NB100-105: HIF-1 alpha Antibody (H1alpha67)
- 2.NB100-134: HIF-1 alpha Antibody
- 3.NB100-296: HIF-1 alpha Antibody (HA111)
- 4.NB100-449: HIF-1 alpha Antibody
- 5.NB100-479: HIF-1 alpha Antibody
HIF-1 alpha specifically detects HIF-1 alpha in Canine, Human, Mouse, Primate, Rabbit, Rat samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.
Specifications
Specifications
| Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) |
| Gene Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp |
| Molecular Weight of Antigen | 93 kDa |
| Target Species | Canine, Human, Rat, Mouse, Rabbit, Rat |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?