missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 241 publication
£419.00
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:2000 - 1:10000, Simple Western 1:200, Flow Cytometry 0.125 μg per 1 million cells in a 150 mcl reaction, ELISA 1:100-1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 μg/mg lysate, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated, Knockdown Validated |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Canine, Chicken, Goat, Primate, Monkey samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated, Knockdown Validated.Specifications
| HIF-1 alpha | |
| Western Blot 1:2000 - 1:10000, Simple Western 1:200, Flow Cytometry 0.125 μg per 1 million cells in a 150 mcl reaction, ELISA 1:100-1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 μg/mg lysate, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| TBS, 0.1% BSA with 0.09% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| HIF1A | |
| IgG | |
| Affinity Purified | |
| Monkey (COS-7) and Rat (review feedback). Has been shown to detect HIF-1 alpha in both mouse and human tissue by IHC with Citrate buffer antigen retrieval. Based on 100% sequence identity, this antibody is expected to react with: Cavia porcellus, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Grass carp, Northern pike, Atlantic cod, Duckbill platypus and Gansu zokor. |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q16665 | |
| 3091 | |
| The immunogen recognized by this HIF-1 alpha Antibody maps to a region between residues 775 and the C-terminus (residue 826) of human hypoxia-inducible factor 1 [Uniprot# Q16665]. | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| 93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title