missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody (H1alpha67), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-105R
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Feline, Ferret, Primate, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated, Knockdown Validated.
Specifications
| HIF-1 alpha | |
| Monoclonal | |
| DyLight 550 | |
| Q16665 | |
| HIF1A | |
| This HIF-1 alpha Antibody (H1alpha67) was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reactivity with Candida albicans reported by a customer review. Reactivity with Xenopus reported in the scientific literature (PMID: 18303027). | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown | |
| H1alpha67 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated, Knockdown Validated | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1 | |
| Mouse | |
| 93 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Feline, Ferret, Primate, Monkey, Rabbit, Sheep, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction