missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody, DyLight 650 - Exon 9, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47178C
This item is not returnable.
View return policy
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot.
Specifications
| HIF-1 alpha | |
| Polyclonal | |
| Western Blot | |
| Q16665 | |
| HIF1A | |
| This HIF-1 alpha Antibody - Exon 9 was developed against a synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within amino acids 350 - 400) [Uniprot# Q16665]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| HIF-1 alpha Antibody - Exon 9 is specific for Exon 9 of HIF-1 alpha. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| Rabbit | |
| 93 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction