missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 389 publications
£248.00 - £493.00
Specifications
| Antigen | HIF-1 alpha |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2ug/ml, Simple Western, Flow Cytometry 1:10 - 1:1000. Use reported in scientific literature (PMID 21917971), Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Immunohistochemistry-Frozen 5 - 10 ug/mL, Immunoblotting reported in scientific literature (PMID 28506759), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 27148974), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18775493
|
Novus Biologicals
NB100-479SS |
0.025 mL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18766473
|
Novus Biologicals
NB100-479 |
0.1 mL |
£493.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Canine, Fish, Goat, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.Specifications
| HIF-1 alpha | |
| Western Blot 1-2ug/ml, Simple Western, Flow Cytometry 1:10 - 1:1000. Use reported in scientific literature (PMID 21917971), Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Immunohistochemistry-Frozen 5 - 10 ug/mL, Immunoblotting reported in scientific literature (PMID 28506759), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 27148974), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Canine, Fish, Goat, Hamster, Primate, Rabbit | |
| Q61221 | |
| 3091 | |
| This HIF-1 alpha Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221]. | |
| Primary | |
| Store at -20°C. | |
| 93 kDa |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
| HIF1A | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat, goat, primate and rabbit. Hamster reactivity reported in scientific literature (PMID: 24997364 and 24997360). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title