missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIF-1 alpha Monoclonal antibody specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-Out
Specifications
Specifications
| Antigen | HIF-1 alpha |
| Applications | Western Blot, Immunocytochemistry, Gene Knock-Out |
| Classification | Monoclonal |
| Clone | 2443B |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 |
| Host Species | Rabbit |
| Immunogen | E. coli-derived recombinant human HIF-1 alpha/HIF1A, Arg575-Asn826, Accession # Q16665.1 |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?